AC5638472953 Accela ChemBio Inc.

Diethyl Iminodicarboxylate

  • MDL number MFCD00031532
  • Cas Number : 19617-44-8
  • Molecular Weight : 161.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form White to yellow solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY046349&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY046349.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart