AC5947205062 Accela ChemBio Inc.

(R)-3-Amino-3-phenyl-1-propanol

  • MDL number MFCD01311791
  • Cas Number : 170564-98-4
  • Molecular Weight : 151.20562

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form liquid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY002040&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY002040.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart