AC5947205308 Accela ChemBio Inc.

Chromium(III) Acetylacetonate

  • MDL number MFCD00000015
  • Cas Number : 21679-31-2
  • Molecular Weight : 349.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY010217&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY010217.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart