AC5947205309 Accela ChemBio Inc.

Bis(cyclopentadienyl)titanium(IV) Dichloride

  • MDL number MFCD00003723
  • Cas Number : 1271-19-8
  • Molecular Weight : 248.96

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. +4?
Physical Form solid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY010218&attname=GoodsCode

Safety Information

signalword Warning
pcodes P261
hcodes GHS07
MSDS http://pdf.accelachem.com/cn/msds/SY010218.pdf
hstatements H315-H335
Availability details for
Quantity
  • Availability
  • Add to Cart