AC5947205486 Accela ChemBio Inc.

3-Amino-3-(3-fluorophenyl)propanoic Acid

  • MDL number MFCD00764457
  • Cas Number : 117391-51-2
  • Molecular Weight : 183.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY014252&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY014252.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart