AC5947205817 Accela ChemBio Inc.

Dimethyl 7,7'-Dimethoxy-[4,4'-bi-1,3-benzodioxole]-5,5'-dicarboxylate

  • MDL number MFCD01751431
  • Cas Number : 73536-69-3
  • Molecular Weight : 418.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form solid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY021274&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY021274.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart