AC5947206032 Accela ChemBio Inc.

6-Amino-2-thiouracil

  • MDL number MFCD00052384
  • Cas Number : 1004-40-6
  • Molecular Weight : 143.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY025189&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY025189.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart