AC5947206116 Accela ChemBio Inc.

2-Amino-2-(3,4-dichlorophenyl)acetic Acid

  • MDL number MFCD03425231
  • Cas Number : 128349-50-8
  • Molecular Weight : 220.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form White to yellow solid
Shipping Temperature RT
Country of Origin CN
Product Link http://www.accelachem.com/cn/search.html?keyword=SY028338&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY028338.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart