AC5947206119 Accela ChemBio Inc.

2-(Cbz-amino)-2-methylpropanamide

  • MDL number MFCD25967457
  • Cas Number : 81587-18-0
  • Molecular Weight : 236.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY028362&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY028362.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart