AC5947206120 Accela ChemBio Inc.

N-[3-(Dimethylamino)propyl]acetamide

  • MDL number MFCD01195313
  • Cas Number : 3197-19-1
  • Molecular Weight : 144.21474

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form liquid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY028365&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY028365.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart