AC5947206176 Accela ChemBio Inc.

3-(Bromomethyl)piperidine Hydrochloride

  • MDL number MFCD21098325
  • Cas Number : 1353976-07-4
  • Molecular Weight : 214.53

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-(bromomethyl)piperidine hydrochloride
Inchi Code 1S/C6H12BrN.ClH/c7-4-6-2-1-3-8-5-6;/h6,8H,1-5H2;1H
Inchi Key MPWJVOIHKVBVED-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. +4?
Physical Form White to yellow solid
Shipping Temperature RT
Country of Origin CN
Product Link http://www.accelachem.com/cn/search.html?keyword=SY029332&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY029332.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart