AC5947206332 Accela ChemBio Inc.

4-Amino-1-methylpyrazole-3-carboxylic Acid

  • MDL number MFCD08558492
  • Cas Number : 1006334-34-4
  • Molecular Weight : 141.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY030212&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY030212.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart