AC5947206446 Accela ChemBio Inc.

Methyl 1-Benzylimidazole-2-carboxylate

  • MDL number MFCD26680731
  • Cas Number : 1502811-19-9
  • Molecular Weight : 216.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY030659&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY030659.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart