AC5947206498 Accela ChemBio Inc.

4,5-Dichloro-2-fluorobenzoic Acid

  • MDL number MFCD00672988
  • Cas Number : 289039-49-2
  • Molecular Weight : 209

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY030903&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY030903.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart