AC5947206548 Accela ChemBio Inc.

Methyl 3-Amino-4-(benzylamino)benzoate

  • MDL number MFCD13343839
  • Cas Number : 68502-22-7
  • Molecular Weight : 256.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY031104&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY031104.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart