AC5947206573 Accela ChemBio Inc.

4-(Bromomethyl)quinoline

  • MDL number MFCD09953523
  • Cas Number : 5632-16-6
  • Molecular Weight : 222.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -20?
Physical Form solid
Shipping Temperature +4 ?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY031349&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY031349.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart