AC5947206592 Accela ChemBio Inc.

2,6-Dibromobenzyl Acetate

  • MDL number MFCD20482187
  • Cas Number : 1147858-83-0
  • Molecular Weight : 307.97

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY031426&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY031426.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart