AC5947206658 Accela ChemBio Inc.

4-Fluoro-2-[(4-methyl-1-piperazinyl)methyl]phenylboronic Acid Pinacol Ester

  • MDL number MFCD28133362
  • Cas Number : 2096340-29-1
  • Molecular Weight : 334.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-(5-fluoro-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzyl)-4-methylpiperazine
Inchi Code 1S/C18H28BFN2O2/c1-17(2)18(3,4)24-19(23-17)16-7-6-15(20)12-14(16)13-22-10-8-21(5)9-11-22/h6-7,12H,8-11,13H2,1-5H3
Inchi Key MWUIDDSJZGGFHS-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. +4?
Physical Form White to yellow solid
Shipping Temperature RT
Country of Origin CN
Product Link http://www.accelachem.com/cn/search.html?keyword=SY031696&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY031696.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart