AC5947206732 Accela ChemBio Inc.

8-Phenyloctanoic Acid

  • MDL number MFCD00037802
  • Cas Number : 26547-51-3
  • Molecular Weight : 220.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY032391&attname=GoodsCode

Safety Information

signalword Warning
pcodes P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
hcodes GHS07
MSDS http://pdf.accelachem.com/cn/msds/SY032391.pdf
hstatements H315-H319
Availability details for
Quantity
  • Availability
  • Add to Cart