AC5947206748 Accela ChemBio Inc.

7-Bromo-2-chloro-5H-pyrrolo[3,2-d]pyrimidine

  • MDL number MFCD22398085
  • Cas Number : 1429882-36-9
  • Molecular Weight : 232.47

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY032484&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY032484.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart