AC5947206749 Accela ChemBio Inc.

Methyl 3,3,3-Trifluoro-2-phenylpropionate

  • MDL number MFCD16617025
  • Cas Number : 184414-29-7
  • Molecular Weight : 218.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form liquid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY032500&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY032500.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart