AC5947206753 Accela ChemBio Inc.

N-(p-Toluenesulfonyl)glycine Methyl Ester

  • MDL number MFCD00558890
  • Cas Number : 2645-02-5
  • Molecular Weight : 243.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY032562&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY032562.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart