AC5947206793 Accela ChemBio Inc.

4-Amino-4'-hydroxybenzophenone

  • MDL number MFCD02648435
  • Cas Number : 14963-34-9
  • Molecular Weight : 213.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY032771&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY032771.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart