AC5947206876 Accela ChemBio Inc.

3-Cyclopropylaniline

  • MDL number MFCD06802404
  • Cas Number : 485402-64-0
  • Molecular Weight : 133.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form Colorless to brown liquid
Shipping Temperature RT
Country of Origin CN
Product Link http://www.accelachem.com/cn/search.html?keyword=SY033396&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY033396.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart