AC5947206891 Accela ChemBio Inc.

N-[4-Cyano-3-(trifluoromethyl)phenyl]-2-methyloxirane-2-carboxamide

  • MDL number MFCD08460201
  • Cas Number : 90357-51-0
  • Molecular Weight : 270.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY033473&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY033473.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart