AC5947206929 Accela ChemBio Inc.

3-Azaspiro[5.5]undecane

  • MDL number MFCD00011572
  • Cas Number : 180-44-9
  • Molecular Weight : 153.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-azaspiro[5.5]undecane
Inchi Code 1S/C10H19N/c1-2-4-10(5-3-1)6-8-11-9-7-10/h11H,1-9H2
Inchi Key LIZKZVQBLDHKCY-UHFFFAOYSA-N

Description

Purity 98%
Storage temp. RT
Physical Form Colorless to yellow liquid
Shipping Temperature RT
Country of Origin CN
Product Link http://www.accelachem.com/cn/search.html?keyword=SY033626&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY033626.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart