AC5947206960 Accela ChemBio Inc.

1,2-Di-p-tolylethylenediamine

  • MDL number MFCD06658956
  • Cas Number : 124842-96-2
  • Molecular Weight : 240.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form solid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY033697&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY033697.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart