AC5947207050 Accela ChemBio Inc.

6-Bromo-2-(chloromethyl)quinazoline

  • MDL number MFCD28977356
  • Cas Number : 1936538-67-8
  • Molecular Weight : 257.51434

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY034019&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY034019.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart