AC5947207114 Accela ChemBio Inc.

5-Methoxypyrimidine

  • MDL number MFCD04971313
  • Cas Number : 31458-33-0
  • Molecular Weight : 110.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 5-methoxypyrimidine
Inchi Code 1S/C5H6N2O/c1-8-5-2-6-4-7-3-5/h2-4H,1H3
Inchi Key JIADELSANNMYFC-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. +4?
Physical Form White to brown solid
Shipping Temperature RT
Country of Origin CN
Product Link http://www.accelachem.com/cn/search.html?keyword=SY034303&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY034303.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart