AC5947207149 Accela ChemBio Inc.

4-(3-Bromopropyl)morpholine Hydrobromide

  • MDL number MFCD13176498
  • Cas Number : 88806-06-8
  • Molecular Weight : 289.01

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form White to yellow solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY034420&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY034420.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart