AC5947207150 Accela ChemBio Inc.

1-(3-Bromopropyl)piperidine Hydrobromide

  • MDL number MFCD13176495
  • Cas Number : 58689-34-2
  • Molecular Weight : 287.04

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form White to yellow solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY034421&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY034421.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart