AC5947207315 Accela ChemBio Inc.

1-(Aminomethyl)isoquinoline

  • MDL number MFCD06213370
  • Cas Number : 40615-08-5
  • Molecular Weight : 158.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form liquid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY035455&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY035455.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart