AC5947207319 Accela ChemBio Inc.

3-(Aminomethyl)-5-bromo-2-methylpyridine

  • MDL number MFCD18255107
  • Cas Number : 1211589-61-5
  • Molecular Weight : 201.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form Colorless to brown liquid, or white to brown solid
Shipping Temperature RT
Country of Origin CN
Product Link http://www.accelachem.com/cn/search.html?keyword=SY035464&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY035464.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart