AC5947207359 Accela ChemBio Inc.

Ethyl 2,4,6-Trichloronicotinate

  • MDL number MFCD28503376
  • Cas Number : 1934657-78-9
  • Molecular Weight : 254.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. +4?
Physical Form liquid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY035696&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY035696.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart