AC5947207401 Accela ChemBio Inc.

3,7-Dibromo-2,6-dihydroxynaphthalene

  • MDL number MFCD28954613
  • Cas Number : 1227743-83-0
  • Molecular Weight : 317.96

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY035972&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY035972.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart