AC5947207449 Accela ChemBio Inc.

3-(4-Chlorophenyl)-2-hydroxypropionic Acid

  • MDL number MFCD11935789
  • Cas Number : 23434-95-9
  • Molecular Weight : 200.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY036070&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY036070.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart