AC5947207510 Accela ChemBio Inc.

3-Ethylthiophene

  • MDL number MFCD00070514
  • Cas Number : 1795-01-3
  • Molecular Weight : 112.19272

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. +4?
Physical Form Colorless to yellow liquid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY036406&attname=GoodsCode

Safety Information

signalword Warning
pcodes P210-P233-P241-P280-P303+P361+P353-P403+P235-P501
hcodes GHS02
MSDS http://pdf.accelachem.com/cn/msds/SY036406.pdf
hstatements H226
Availability details for
Quantity
  • Availability
  • Add to Cart