AC5947207723 Accela ChemBio Inc.

Ethyl 2-(4-Methyl-5-thiazolyl)acetate

  • MDL number MFCD06410796
  • Cas Number : 406727-23-9
  • Molecular Weight : 185.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form liquid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY037196&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY037196.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart