AC5947207833 Accela ChemBio Inc.

(2R,3R,3aS,9aR)-3-Hydroxy-2-(hydroxymethyl)-7-methyl-3,3a-dihydro-2H-furo[2',3':4,5]oxazolo[3,2-a]pyrimidin-6(9aH)-one

  • MDL number MFCD00233555
  • Cas Number : 22423-26-3
  • Molecular Weight : 240.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY038115&attname=GoodsCode

Safety Information

hcodes no data known
MSDS http://pdf.accelachem.com/cn/msds/SY038115.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart