AC5947207911 Accela ChemBio Inc.

(3aR,3'aR,4R,4'R,6R,6'R,7aS,7'aS)-3a,3'a,5,5,5',5'-Hexamethyldodecahydro-2,2'-bi[4,6-methanobenzo[d][1,3,2]dioxaborole]

  • MDL number MFCD02683578
  • Cas Number : 230299-17-9
  • Molecular Weight : 358.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY038439&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY038439.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart