AC5947207937 Accela ChemBio Inc.

2,2-Difluorocyclopentane-1,3-dione

  • MDL number MFCD16657105
  • Cas Number : 1215071-21-8
  • Molecular Weight : 134.0808664

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY038555&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY038555.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart