AC5947207979 Accela ChemBio Inc.

2'-Bromo-5'-methoxyacetanilide

  • MDL number MFCD12923200
  • Cas Number : 123027-99-6
  • Molecular Weight : 244.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY038711&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY038711.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart