AC5947208013 Accela ChemBio Inc.

alpha,alpha-Difluoro-2-(4-methoxyphenyl)acetic Acid

  • MDL number MFCD11007700
  • Cas Number : 1027513-97-8
  • Molecular Weight : 202.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY038857&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY038857.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart