AC5947208022 Accela ChemBio Inc.

Ethyl 2,2-Difluoro-2-(4-nitrophenyl)acetate

  • MDL number MFCD11975616
  • Cas Number : 243656-25-9
  • Molecular Weight : 245.1795664

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. RT
Physical Form liquid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY038866&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY038866.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart