AC5947208202 Accela ChemBio Inc.

(R)-4-(Dibenzylamino)-3-fluoro-2-methyl-2-butanol

  • MDL number MFCD29055296
  • Cas Number : 1609545-83-6
  • Molecular Weight : 301.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form solid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY039317&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY039317.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart