AC5947208268 Accela ChemBio Inc.

2-Amino-6-bromo-3H-imidazo[4,5-b]pyridine

  • MDL number MFCD16845543
  • Cas Number : 1250115-39-9
  • Molecular Weight : 213.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY039629&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY039629.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart