AC5947208300 Accela ChemBio Inc.

4-Cyano-3-(trifluoromethyl)phenyl Isothiocyanate

  • MDL number MFCD09800709
  • Cas Number : 143782-23-4
  • Molecular Weight : 228.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. +4?
Physical Form White to yellow solid
Shipping Temperature RT
Country of Origin CN
Product Link http://www.accelachem.com/cn/search.html?keyword=SY039781&attname=GoodsCode

Safety Information

signalword Warning
pcodes P261-P264-P270-P271-P280-P301+P312+P330-P302+P352+P312+P362+P364-P304+P340+P312
MSDS http://pdf.accelachem.com/cn/msds/SY039781.pdf
pictograms GHS07
hstatements H302+H312+H332-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart