AC5947208320 Accela ChemBio Inc.

3-Amino-6-bromoimidazo[1,2-a]pyridine

  • MDL number MFCD09864901
  • Cas Number : 705262-55-1
  • Molecular Weight : 212.04664

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY039838&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY039838.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart