AC5947208401 Accela ChemBio Inc.

2-[4-(4-Chloro-2-fluorophenyl)-1-piperazinyl]aniline

  • MDL number MFCD29923990
  • Cas Number : 2006277-40-1
  • Molecular Weight : 305.78

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form solid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY040078&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY040078.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart