AC5947208522 Accela ChemBio Inc.

1-[5-(Trifluoromethyl)-2-pyridyl]-1-propanol

  • MDL number MFCD30342410
  • Cas Number : 1595889-36-3
  • Molecular Weight : 205.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form liquid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY041301&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY041301.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart